Learn More
HOXC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541479
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KIWFQNRRMK WKKESNLTST LSGGGGGATA DSLGGKEEKR EETEEEKQKE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Sheep: 100%
HOXC6 Polyclonal antibody specifically detects HOXC6 in Mouse samples. It is validated for Immunohistochemistry.Specifications
HOXC6 | |
Polyclonal | |
Unconjugated | |
HOXC6 | |
Antennapedia-type homeotic factor; CP25; HHO.C8; homeo box 3C; homeo box C6; homeo box C8 protein; homeobox C6; homeobox protein CP25; Homeobox protein HHO.C8; homeobox protein Hox-3.3; Homeobox protein Hox-3C; homeobox protein Hox-6.1; Homeobox protein Hox-C6; Hox C6; HOX3; Hox-3.3; HOX3C; Hox-6.1; Hoxc6; Hoxc-6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15425 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P10629 | |
HOXC6 | |
Synthetic peptide directed towards the C-terminal of human HOXC6 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |