Learn More
homeobox C12, Mouse, Clone: 1C6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HOXC12.
Brand: Abnova H00003228-M01.100ug
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. [provided by RefSeq
Sequence: MGEHNLLNPGFVGPLVNIHTGDTFYFPNFRASGAQLPGLPSLSYPRRDNVCSLSWPSAEPCNGYPQPYLGSPVSLNPPFGRTCELARVEDGKGYYREPCASpecifications
| homeobox C12 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_173860 | |
| HOXC12 | |
| HOXC12 (NP_776272, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 1C6 | |
| Mouse monoclonal antibody raised against a partial recombinant HOXC12. | |
| HOXC12 | |
| HOC3F/HOX3/HOX3F | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3228 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.