Learn More
HOXB5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535898
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: HepG2 WCE and NE. Predicted reactivity: Mouse (97%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HOXB5 Polyclonal antibody specifically detects HOXB5 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
HOXB5 | |
Polyclonal | |
Unconjugated | |
HOXB5 | |
AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P09067 | |
HOXB5 | |
Recombinant fragment corresponding to a region within amino acids 18 and 205 of HOXB5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |