Learn More
hnRNP H1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527610
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, Raji, NCI-H929, mouse brain. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (87%), Xenopus laevis (91%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some may shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNA and is very similar to the family member HNRPF. This gene may be associated with hereditary lymphedema type I. Alternatively spliced transcript variants have been described.Specifications
hnRNP H1 | |
Polyclonal | |
Unconjugated | |
Hnrnph1 | |
AI642080; E430005G16Rik; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); hnRNP H; Hnrnph; HNRNPH1; HNRPH; HNRPH1; Ratsg1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3187, 59013 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O35737, P31943 | |
Hnrnph1 | |
Recombinant fragment corresponding to a region within amino acids 50 and 289 of Human hnRNP H | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |