missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGA2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519197
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
HMGA2, previously known as HMGI-C, belongs to the group of high mobility chromosomal proteins that have the AT-hook DNA-binding motif. It is highly and ubiquitously expressed during embryogenesis. Chromosomal rearrangements of the HMGA2 gene and overexpression are frequently associated with benign and malignant tumors, respectively. Human and mouse HMGA2 shares 95% amino acid sequence identity.
Specifications
| HMGA2 | |
| Polyclonal | |
| Unconjugated | |
| Hmga2 | |
| 9430083A20Rik; BABL; high mobility group AT-hook 2; high mobility group AT-hook protein 2; High mobility group protein HMGI-C; high mobility group protein I, isoform C; high-mobility group (nonhistone chromosomal) protein isoform I-C; High-mobility group protein HMGI-C; HMGA2; Hmgic; HMGI-C; LIPO; non-histone chromosomal architectural protein HMGI-C; pg; pygmy; STQTL9 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8091 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P52926 | |
| Hmga2 | |
| Synthetic peptide sequence (KAAQKKAEATGEK) corresponding to the internal amino acids of HMGA2 (aa 62-75). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction