Learn More
HLA-DR Mouse anti-Human, PE, Clone: TU36, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555561
Description
Reacts with major histocompatibility Class II HLA-DQ1 and DQ2 antigens and weakly with HLA-DQ3. HLA-DQ is expressed primarily on immunocompetent B cells and also on macrophages. This antibody does not cross-react with HLA-DR2 and DR7. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
| HLA-DR | |
| Monoclonal | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Rabbit, Rhesus Monkey | |
| IgG2b κ |
| Flow Cytometry | |
| TU36 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.