missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse anti-Human, PE, Clone: G46-6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560943

Product Code. 15870619

  • ONLINE EXCLUSIVE PRICE
    £152.00
    £145.76 / Each
    Save £6.24
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The G46-6 monoclonal antibody specifically binds to HLA-DR, a major histocompatibility complex (MHC) class II antigen. HLA-DR antigens are encoded by genes within the Human Leukocyte Antigen (HLA) Complex located on chromosome 6. HLA-DR is a transmembrane heterodimeric glycoprotein composed of an α chain (36 kDa) and a β subunit (27 kDa) expressed primarily on antigen presenting cells: B cells, dendritic cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in mediating cellular interactions during antigen presentation to CD4-positive T cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-DR
Monoclonal
PE
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
G46-6
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.