Learn More
HLA-DR Mouse anti-Human, PE, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556644
Description
Clone G46-6 cross-reacts with the human class II antigen of the major histocompatability complex (MHC). This clone also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on lymphocytes is similar to that observed with human peripheral blood lymphocytes, with the majority of CD20-positive cells and a small subset of CD3-positive cells being positive with G46-6. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-6
Isotype: IgG2a κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
| HLA-DR | |
| Monoclonal | |
| PE | |
| Mouse | |
| 50 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Rhesus Monkey | |
| IgG2a κ |
| Flow Cytometry | |
| L243 (G46-6) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.