Learn More
major histocompatibility complex, class II, DO alpha (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HLA-DOA protein.
Brand: Abnova H00003111-B01.50uL
Description
HLA-DOA belongs to the HLA class II alpha chain paralogues. HLA-DOA forms a heterodimer with HLA-DOB. The heterodimer, HLA-DO, is found in lysosomes in B cells and regulates HLA-DM-mediated peptide loading on MHC class II molecules. In comparison with classical HLA class II molecules, this gene exhibits very little sequence variation, especially at the protein level. [provided by RefSeq
Sequence: MALRAGLVLGFHTLMTLLSPQEAGATKADHMGSYGPAFYQSYGASGQFTHEFDEEQLFSVDLKKSEAVWRLPEFGDFARFDPQGGLAGIAAIKAHLDILVERSNRSRAINVPPRVTVLPKSRVELGQPNILICIVDNIFPPVINITWLRNGQTVTEGVAQTSFYSQPDHLFRRFHYLPFVPSAEDVYDCQVEHWGLDAPLLRHWELQVPIPPPDAMETLVCALGLAIGLVGFLVGTVLIIMGTYVSSVPRSpecifications
| major histocompatibility complex, class II, DO alpha | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human HLA-DOA protein. | |
| HLA-DOA | |
| HLA-DNA/HLA-DZA/HLADZ | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC013183 | |
| HLA-DOA | |
| HLA-DOA (AAH13183, 1 a.a. ∼ 250 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 3111 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.