Learn More
anti-HLA Class I, Clone: W6/32, Abcam™
Brand: Abcam AB23755-100UG
Description
Host Species: Mouse
Clone: W6/32
Isotype: IgG2a
Cross Reactivity: Cow, Cat, Human
Immunogen: Tissue / cell preparation - Membrane of human tonsil cells.
Applications:
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting
Specifications
| HLA Class I | |
| W6/32 | |
| Unconjugated | |
| Tissue / cell preparation - Membrane of human tonsil cells. | |
| Immunology | |
| Bovine, Feline, Human | |
| IgG2a |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.