missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533851
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gibbon, Monkey (94%).
The Histamine H4 receptor has been reported in human blood peripheral leukocytes, bone marrow, colon, liver, lung, smalll intestine, spleen, testis, thymus, tonsil, and trachea. Contrary to the mouse H4 receptor, the human gene was not detected in brain. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
| HRH4 | |
| Polyclonal | |
| Unconjugated | |
| HRH4 | |
| AXOR35; BG26; GPCR105; Gprc105; G-protein coupled receptor 105; GPRv53; H4; H4R; HH4R; histamine H4 receptor; histamine receptor; histamine receptor H4; Hrh4; MGC130500; Pfi-013; SP9144 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 59340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9H3N8 | |
| HRH4 | |
| Synthetic 17 amino acid peptide from C-terminus of human HRH4 / Histamine H4 Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction