missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£204.00 - £396.00
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:3500, Simple Western 1:250 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097803
|
Novus Biologicals
NBP1-02160SS |
0.025 mL |
£204.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779583
|
Novus Biologicals
NBP1-02160 |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Simple Western.Specifications
| HIF-1 alpha | |
| Western Blot 1:3500, Simple Western 1:250 | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| HIF1A | |
| IgG | |
| Affinity Purified | |
| Human. Reactivity with mouse and porcine reported by a customer review. Predicted to react with monkey based on 100% sequence homology. Immunogen sequence has 96% homology to bovine, 95% to porcine, 94% to rabbit, 92% to rat, 93% to mouse isoform 2, 83% to mouse isoform 1, and 89% to chicken. |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Pig, Bovine | |
| Q16665 | |
| 3091 | |
| This HIF-1 alpha Antibody was developed against a genomic sequence made to an internal portion of human HIF-1 alpha (within amino acids 400 - 550) [Uniprot# Q16665]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title