missing translation for 'onlineSavingsMsg'
Learn More

anti-Herpes Simplex Virus 2 (HSV2), Polyclonal, Novus Biologicals™

Product Code. 18374071 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
6 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374071 6 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374071 Supplier Novus Biologicals Supplier No. NBP278056JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Progesterone Monoclonal specifically detects Progesterone in All Species samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus 2 (HSV2)
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS
Host Species Rabbit
Immunogen Whole rabbit corneal cells, infected with herpes simplex virus type II (strain M5), were solubilized with detergent.
Quantity 6 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This reacts with Herpes Simplex Virus 2 (HSV2) specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
Target Species Virus
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.