missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSV Type 1 Monoclonal Antibody (LP10)

HSV Type 1 Monoclonal Antibody (LP10)
Brand: Invitrogen MA516285
This item is not returnable.
View return policy
Description
HSV Type 1 Monoclonal Antibody for ICC/IF
Specifications
| HSV Type 1 | |
| Monoclonal | |
| 6.45 mg/mL | |
| PBS with 15mM sodium azide; pH 7.5 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG3 |
| Immunocytochemistry | |
| LP10 | |
| Unconjugated | |
| Herpes; Herpes Simplex; Herpes Simplex 1; Herpes Simplex I; herpes simplex virus 1; Herpes Simplex Virus Type 1; Herpes Virus; HHV; HSV; HSV Type 1; HSV1; KITH_HHV1S | |
| HSV I (strain HFEM). | |
| 2 mg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction