missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSV Type 1 (strain F) Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA17496
This item is not returnable.
View return policy
Description
The PA1-7496 antibody reacts with Herpes Simplex virus 1. PA1-7496 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-7496 antibody was raised against HSV Strain F ICP + virion antigens.
HSV is a member of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.
Specifications
| HSV Type 1 (strain F) | |
| Polyclonal | |
| Biotin | |
| Herpes; Herpes Simplex; Herpes Simplex 1; Herpes Simplex I; herpes simplex virus 1; Herpes Simplex Virus Type 1; Herpes Virus; HHV; HSV; HSV Type 1; HSV1; KITH_HHV1S | |
| HSV Strain F ICP + virion antigens. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 5.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction