Learn More
hect domain and RLD 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HERC3 protein.
Brand: Abnova H00008916-D01P.100ug
Description
HERC3 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).[supplied by OMIM
Sequence: MLCWGYWSLGQPGISTNLQGIVAEPQVCGFISDRSVKEVACGGNHSVFLLEDGEVYTCGLNTKGQLGHEREGNKPEQIGALADQHIIHVACGESHSLALSDRGQLFSWGAGSDGQLGLMTTEDSVAVPRLIQKLNQQTILQVSCGNWHCLALAADGQFFTWGKNSHGQLGLGKEFPSQASPQRVRSLEGIPLAQVAAGGAHSFALSLSGAVFGWGMNNAGQLGLSDEKDRESPCHVKLLRTQKVVYISCGEEHTAVLTKSGGVFTFGAGSCGQLGHDSMNDEVNPRRVLELMGSEVTQIACGRQHTLAFVPSSGLIYAFGCGARGQLGTGHTCNVKCPSPVKGYWAAHSGQLSARADRFKYHIVKQIFSGGDQTFVLCSKYENYSPAVDFRTMNQAHYTSLINDETIAVWRQKLSEHNNANTINGVVQILSSAACWNGSFLEKKIDEHFKTSPKIPGIDLNSTRVLFEKLMNSQHSMILEQILNSFESCLIPQLSSSPPDVEAMRIYLILPEFPLLQDSKYYITLTIPLAMAILRLDTNPSKVLDNWWSQVCPKYFMKLVNLYKGAVLYLLRGRKTFLIPVLFNNYITAALKLLEKLYKVNLKVKHVEYDTFYIPEISNLVDIQEDYLMWFLHQAGMKARPSIIQDTVTLCSYPFIFDAQAKTKMLQTDAELQMQVAVNGANLQNVFMLLTLEPLLARSPFLVLHVRRNNLVGDALRELSIHSDIDLKKPLKVIFDGEEAVDAGGVTKEFFLLLLKELLNPIYGMFTYYQDSNLLWFSDTCFVEHNWFHLIGITCGLAIYNSTVVDLHFPLALYKKLLNVKPGLEDLKELSPTEGRSLQELLDYPGEDVEETFCLNFTICRESYGVIEQKKLIPGGDNVTVCKDNRQEFVDAYVNYVFQISVHEWYTAFSSGFLKVCGGKVLELFQPSELRAMMVGNSNYNWEELEETAIYKGDYSATHPTVKLFWETFHEFPLEKKKKFLLFLTGSDRIPIYGMASLQIVIQSTASGEEYLPVAHTCYNLLDLPKYSSKEILSARLTQALDNYEGFSLASpecifications
| hect domain and RLD 3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human HERC3 protein. | |
| HERC3 | |
| KIAA0032 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014606.1 | |
| HERC3 | |
| HERC3 (NP_055421.1, 1 a.a. ∼ 1050 a.a) full-length human protein. | |
| 100 μg | |
| Ubiquitin/Proteasome | |
| 8916 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.