missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2) Monoclonal Antibody (e2-4001), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513102
This item is not returnable.
View return policy
Description
MA5-13102 targets HER-2 in IHC, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13102 immunogen is cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein.
ErbB2 (HER-2) Monoclonal antibody specifically detects ErbB2 (HER-2) in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.
Specifications
| ErbB2 (HER-2) | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P04626, P70424 | |
| ERBB2 | |
| Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| e2-4001 | |
| Biotin | |
| ERBB2 | |
| avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
| Mouse | |
| Protein G | |
| RUO | |
| 13866, 2064 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction