missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMOX1 Monoclonal Antibody (GTS-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125475
This item is not returnable.
View return policy
Description
MA1-25475 will not react with rabbit.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
Specifications
| HMOX1 | |
| Monoclonal | |
| 2 mg/mL | |
| 10mM PBS with 1% BSA and no preservative; pH 7.4 | |
| P06762, P09601, P14901 | |
| HMOX1 | |
| Rat heme oxygenase 1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| GTS-1 | |
| Unconjugated | |
| HMOX1 | |
| bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein | |
| Mouse | |
| purified | |
| RUO | |
| 15368, 24451, 3162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction