Learn More
helicase with zinc finger, Mouse, Clone: 5B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HELZ.
Brand: Abnova H00009931-M02.100ug
Description
HELZ is a member of the superfamily I class of RNA helicases. RNA helicases alter the conformation of RNA by unwinding double-stranded regions, thereby altering the biologic activity of the RNA molecule and regulating access to other proteins (Wagner et al., 1999 [PubMed 10471385]).[supplied by OMIM
Sequence: MEDRRAEKSCEQACESLKRQDYEMALKHCTEALLSLGQYSMADFTGPCPLEIERIKIESLLYRIASFLQLKNYVQADEDCRHVLGEGLAKGEDAFRAVLCSpecifications
| helicase with zinc finger | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014877 | |
| HELZ | |
| HELZ (NP_055692, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 9931 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 5B2 | |
| Mouse monoclonal antibody raised against a partial recombinant HELZ. | |
| HELZ | |
| DHRC/DKFZp586G1924/DRHC/HUMORF5/KIAA0054/MGC163454 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Safety and Handling
- HELZ monoclonal antibody (M02), clone 5B2
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium chloride, sodium phosphate dibasic, potassium phosphate monobasic, potassium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.