missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter pylori Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516813
This item is not returnable.
View return policy
Description
PA5-16813 targets Helicobacter pylori in IHC (P) applications and shows reactivity with Bacteria samples. The PA5-16813 immunogen is purified Helicobacter pylori.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.Specifications
Helicobacter pylori | |
Polyclonal | |
Unconjugated | |
H. pylori; Campylobacter pyloridis | |
Purified Helicobacter pylori | |
500 μL | |
Primary | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |