Learn More
ITIH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA11756
Descripción
PA1-1756 detects HC2 from human and mouse samples. PA1-1756 has been successfully used in immunohistochemical staining of paraffin embedded tissue sections. The PA1-1756 immunogen is a synthetic peptide corresponding to residues E A N N L G L L D P N. This immunizing peptide (PEP-302) is available for neutralization and control experiments.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Especificaciones
| ITIH2 | |
| Polyclonal | |
| Unconjugated | |
| ITIH2 | |
| PSMA1, proteasome (prosome, macropain) subunit, alpha type, 1, 30 kDa prosomal protein, Macropain subunit C2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16425, 3698 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P19823, Q61703 | |
| ITIH2 | |
| Synthetic peptide corresponding to residues E A N N L G L L D P N. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.