Learn More
HBZ, Mouse, Clone: 3C4-1D5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant HBZ.
Brand: Abnova H00003050-M01.100ug
Description
Zeta-globin is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that includes five functional genes and two pseudogenes. The order of genes is: 5' - zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 -alpha-1 - theta1 - 3'. [provided by RefSeq]
Sequence: MSLTKTERTIIVSMWAKISTQADTIGTETLERLFLSHPQTKTYFPHFDLHPGSAQLRAHGSKVVAAVGDAVKSIDDIGGALSKLSELHAYILRVDPVNFKLLSHCLLVTLAARFPADFTAEAHAAWDKFLSVVSSVLTEKYRSpecifications
| HBZ | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC027892 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3050 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 3C4-1D5 | |
| Mouse monoclonal antibody raised against a full length recombinant HBZ. | |
| HBZ | |
| HBZ | |
| HBZ (AAH27892, 1 a.a. ∼ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.