missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | HBcAg |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin Neat, Immunohistochemistry-Frozen Neat |
| Formulation | PBS with Sodium Azide |
| Gene Alias | c, Core antigen, HBc, HBcAg, Hepatitis B Virus core antigen |
| Gene Symbols | HBVgp4 |
| Host Species | Rabbit |
| Immunogen | HBcAg purified from the lysate of E.coli infected with the viral core DNA. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?