Learn More
HAP1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HAP1 protein.
Brand: Abnova H00009001-D01P.100ug
Description
Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. This gene encodes a protein that interacts with huntingtin, with two cytoskeletal proteins (dynactin and pericentriolar autoantigen protein 1), and with a hepatocyte growth factor-regulated tyrosine kinase substrate. The interactions with cytoskeletal proteins and a kinase substrate suggest a role for this protein in vesicular trafficking or organelle transport. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MRPKRLGRCCAGSRLGPGDPAALTCAPSPSASPAPEPSAQPQARGTGQRVGSRATSGSQFLSEARTGARPASEAGAKAGARRPSAFSAIQGDVRSMPDNSDAPWTRFVFQGPFGSRATGRGTGKAAGIWKTPAAYVGRRPGVSGPERAAFIRELEEALCPNLPPPVKKITQEDVKVMLYLLEELLPPVWESVTYGMVLQRERDLNTAARIGQSLVKQNSVLMEENSKLEALLGSAKEEILYLRHQVNLRDELLQLYSDSDEEDEDEEEEEEEKEAEEEQEEEEAEEDLQCAHPCDAPKLISQEALLHQHHCPQLEALQEKLRLLEEENHQLREEASQLDTLEDEEQMLILECVEQFSEASQQMAELSEVLVLRLENYERQQQEVARLQAQVLKLQQRCRMYGAETEKLQKQLASEKEIQMQLQEESVWVGSQLQDLREKYMDCGGMLIEMQEEVKTLRQQPPVSTGSATHYPYSVPLETLPGFQETLAEELRTSLRRMISDPVYFMERNYEMPRGDTSSLRYDFRYSEDREQVRGFEAEEGLMLAADIMRGEDFTPAEEFVPQEELGAAKKVPAEEGVMEEAELVSEETEGWEEVELELDEATRMNVVTSALEASGLGPSHLDMNYVLQQLANWQDAHYRRQLRWKMLQKGECPHGALPAASRTSCRSSCRSpecifications
| HAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human HAP1 protein. | |
| HAP1 | |
| HAP2/HIP5/HLP/hHLP1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9001 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC156728.1 | |
| HAP1 | |
| HAP1 (AAI56729.1, 1 a.a. ∼ 671 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.