missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAGH, Mouse, Clone: 4D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HAGH.
Brand: Abnova H00003029-M01.100ug
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. [provided by RefSeq
Sequence: GRLPPDTRVYCGHEYTINNLKFARHVEPGNAAIREKLAWAKEKYSIGEPTVPSTLAEEFTYNPFMRVREKTVQQHAGETDPVTTMRAVRREKDQFKMPRDSpecifications
| HAGH | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005326 | |
| HAGH | |
| HAGH (NP_005317, 161 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 4D3 | |
| Mouse monoclonal antibody raised against a partial recombinant HAGH. | |
| HAGH | |
| GLO2/GLX2/GLXII/HAGH1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3029 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction