Learn More
H-2K[k] Mouse anti-Mouse, FITC, Clone: 36-7-5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553592
Description
The 36-7-5 antibody reacts with the H-2K[k] MHC class I alloantigen. Cross-reactivity with splenocytes of SJL/Hsd mice has not been observed by flow cytometric analysis. Reactivity with other haplotypes (e.g., b, d, q) has not been observed. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 36-7-5
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: A.AL mouse splenocytes
Flow Cytometry
Specifications
| H-2K[k] | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| A.AL mouse splenocytes | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 36-7-5 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.