Learn More
H-2K[d] Mouse anti-Mouse, Biotin, Clone: SF1-1.1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553564
Description
The SF1-1.1 antibody reacts with the α3 domain of the H-2K[d] MHC class I alloantigen. Reactivity with other haplotypes (e.g, b, j, k, p, q, s, v) has not been observed. It has been reported that plate-bound SF1-1.1 mAb moderately enhances the apopt
Host Species: Mouse
Clone: SF1-1.1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB/c mouse cells
Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| H-2K[d] | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB/c mouse cells | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| SF1-1.1 | |
| Biotin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.