missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (H+L) Mouse anti-Guinea Pig, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA180575
This item is not returnable.
View return policy
Description
Mouse anti Guinea pig IgG antibody, clone MsGp3 recognizes guinea pig immunoglobulin γ heavy chain and reacts with a subset of B lymphocytes in lymph nodes and is restricted to mantle zone B cells of the follicles.
Thermo Scientific Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Guinea Pig IgG (H+L) | |
| Monoclonal | |
| Unconjugated | |
| EN-L2C guinea pig leukemia B cell line. | |
| RUO | |
| Guinea Pig | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen) | |
| MsGp3 | |
| Mouse | |
| 2 mL | |
| Secondary | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction