Learn More
guanylate cyclase 2D, membrane (retina-specific), Mouse, Clone: 6D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GUCY2D.
Brand: Abnova H00003000-M02.100ug
Beskrivning
This gene encodes a retina-specific guanylate cyclase, which is a member of the membrane guanylyl cyclase family. Like other membrane guanylyl cyclases, this enzyme has a hydrophobic amino-terminal signal sequence followed by a large extracellular domain, a single membrane spanning domain, a kinase homology domain, and a guanylyl cyclase catalytic domain. In contrast to other membrane guanylyl cyclases, this enzyme is not activated by natriuretic peptides. Mutations in this gene result in Leber congenital amaurosis and cone-rod dystrophy-6 diseases. [provided by RefSeq
Sequence: RKVAQGSRSSLGARSMSDIRSGPSQHLDSPNIGVYEGDRVWLKKFPGDQHIAIRPATKTAFSKLQELRHENVALYLGLFLARGAEGPAALWEGNLAVVSEHCTRGSLQDLSpecifikationer
| guanylate cyclase 2D, membrane (retina-specific) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000180 | |
| GUCY2D | |
| GUCY2D (NP_000171, 521 a.a. ∼ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6D9 | |
| Mouse monoclonal antibody raised against a partial recombinant GUCY2D. | |
| GUCY2D | |
| CORD5/CORD6/CYGD/GUC1A4/GUC2D/LCA/LCA1/RETGC-1/ROS-GC1/retGC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3000 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Säkerhet och hantering
- GUCY2D monoclonal antibody (M02), clone 6D9
Signalord
- Warning
Riskkategorier
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Faroangivelser
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Skyddsangivelse
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.