Learn More
GTP binding protein 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GTPBP1.
Brand: Abnova H00009567-A01.50uL
Description
This gene is upregulated by interferon-gamma and encodes a protein that is a member of the AGP11/GTPBP1 family of GTP-binding proteins. A structurally similar protein has been found in mouse, where disruption of the gene for that protein had no observable phenotype. [provided by RefSeq
Sequence: MDEGCGETIYVIGQGSDGTEYGLSEADMEASYATVKSMAEQIEADVILLRERQEAGGRVRDYLVRKRVGDNDFLEVSpecifications
| GTP binding protein 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GTPBP1. | |
| GTPBP1 | |
| GP-1/GP1/HSPC018/MGC20069 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9567 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004286 | |
| GTPBP1 | |
| GTPBP1 (NP_004277, 1 a.a. ∼ 76 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.