Learn More
GTL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540859
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: YGTNYIETLR VQIHANCRIR RVYFSDRLYS EDELPAEFKL YLPVQNKAKQ Sequence homology: Cow: 100%; Dog: 100%; Goat: 77%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
GTL3 Polyclonal antibody specifically detects GTL3 in Human samples. It is validated for Western Blot.Specifications
GTL3 | |
Polyclonal | |
Unconjugated | |
CFAP20 | |
2600014O15Rik; AL024127; Basal body up-regulated protein 22; BUG22; C16orf80; C18H16orf80; C2H16orf80; C3H16orf80; C5H16orf80; CFAP20; cilia and flagella associated protein 20; cilia- and flagella-associated protein 20; EVORF; flagellar associated protein 20 homolog; fSAP23; functional spliceosome-associated protein 23; gene trap locus 3; gene trap locus 3 protein; Gene trap locus 3 protein homolog; GTL3; likely ortholog of mouse gene trap locus 3; similar to transcription factor IIB; T10-2A2; transcription factor 2B; transcription factor IIB; UPF0468 protein C16orf80; UPF0468 protein C16orf80 homolog; zgc:73380 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29105 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y6A4 | |
CFAP20 | |
Synthetic peptide directed towards the C-terminal of human GTL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |