missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GST Tag Recombinant Rabbit Monoclonal Antibody (1H14L28)

Product Code. 15336994
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15336994 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15336994 Supplier Invitrogen™ Supplier No. 700775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

GST (Glutathione S-transferase) is a 26kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GST Tag
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H14L28
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Host Species Rabbit
Immunogen A recombinant protein corresponding to amino acids 1-220.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.