missing translation for 'onlineSavingsMsg'
Learn More

anti-GSK3B, IKBKG Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0300.1Set

Product Code. 16181254

  • £585.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes: GSK3B rabbit purified polyclonal antibody (20μg); IKBKG mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the GSK3B protein, and the other against the IKBKG protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

GSK3B
Monoclonal/Polyclonal
GSK3B
Mouse, Rabbit
RUO
2932/8517
Human
GSK3B & IKBKG Protein Protein Interaction Antibody Pair
In situ PLA
Unconjugated
GSK3B
1 Set
Primary
Antibody pair set content: 1. GSK3B rabbit purified polyclonal antibody (20 ug) 2. IKBKG mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.