Learn More
GSK-3b (pY216) Mouse, Unlabeled, Clone: 13A, BD
Mouse Monoclonal Antibody
£260.82 - £401.97
Specifications
| Antigen | GSK-3b (pY216) |
|---|---|
| Clone | 13A |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15826169
|
BD Biosciences
612312 |
50 μg |
£272.00 £260.82 / Each Save £11.18 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15836169
|
BD Biosciences
612313 |
150 μg |
£419.00 £401.97 / Each Save £17.03 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycogen Synthase Kinase-3β (GSK-3β) is a serine/threonine kinase that affects glycogen metabolism by phosphorylating and down-regulating the activity of muscle glycogen synthase. GSK-3β is identical to the Tau Protein Kinase I (TPK I) that plays a role in the formation of the histopathological brain lesions of Alzheimer's disease (AD). Phosphorylation of the cytoskeletal protein, tau, by GSK-3β converts these proteins into paired helical filaments (PHF) which are found in the neurofibrillary tangles and degenerative neurites of AD patients. Regulation of GSK-3β activity through both serine and tyrosine phosphoylation is a critical determinant of cell death or survival. Factors that promote cell survival, such as growth factors, activate Akt which, in turn phosphorylates GSK-3β at Ser-9, leading to inactivation of its kinase activity. On the contrary, events that promote cell death, such as growth factor removal, cause increases in phosphorylation within the catalytic domain at Tyr-216 and stimulate kinase activity. Thus GSK-3β is a tightly regulated death promoting kinase that regulates the activity of various proteins, including cytokeletal and enzymatic proteins.
Western Blotting
Specifications
| GSK-3b (pY216) | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 13A | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Rat GSK-3β (pY216) Peptide | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.