missing translation for 'onlineSavingsMsg'
Learn More

GS28 Mouse, Unlabeled, Clone: 1, BD

Product Code. 15835829
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835829 50 μg 1 set
15845829 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15835829 Leverantör BD Biosciences Leverantörsnummer 611184

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

Most vesicular fusion events require the function of N-ethylmaleimide-sensitive factor (NSF) and the soluble NSF attachment proteins (SNAPs). Their function is dependent upon the SNAP receptor (SNARE) family of proteins. The specificity of vesicle docking and fusion is mediated by specific interactions between v-SNAREs on vesicles and t-SNAREs on target membranes. GS28 (Golgi SNARE with a size of 28kDa) is a SNARE that associates with the cis-Golgi and participates in trafficking between the ER and the Golgi and between Golgi
compartments. The majority of GS28, the first 230 and 250 aa, is thought to be anchored to the membrane via the C-terminal hydrophobic tail, which is formed by the remaining 20 aa. GS28 and syntaxin 5, another SNARE, exist as a protein complex in the Golgi and this complex is important for the function of both proteins in ER-Golgi transport. The GS28/syntaxin 5 complex can be dissociated by α-SNAP and NSF. In addition, GS28 is thought to interact with α-SNAP when the GS28/syntaxin 5 complex is dissociated. Thus, GS28 is a SNARE protein that mediates, in complex with syntaxin 5, transport within the Golgi and between Golgi and ER.

Host Species: Mouse
Clone: 1
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat GS28 aa. 3-108
Formula Weight [Chemical]: 28kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifikationer

Antigen GS28
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat GS28 aa. 3-108
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Visa mer Visa mindre

For Research Use Only.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.