Learn More
GS15 Mouse, Unlabeled, Clone: 19, BD
Mouse Monoclonal Antibody
£237.00
Specifications
| Antigen | GS15 |
|---|---|
| Clone | 19 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15835729
|
BD Biosciences
610960 |
50 μg | |||||||
Description
Eukaryotic protein trafficking involves the packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the synaptic plasma membrane interact to form a fusion complex which mediates specific docking and fusion of vesicles. This complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs) that include synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosome-associated protein of 25kDa). SNAP-25 and syntaxin are associated with the target plasma membrane (t-SNAREs), while synaptobrevin and synaptotagmin are vesicle-associated proteins (v-SNAREs). In Drosophila, protein transport from the ER to Golgi involves Bet1p, a v-SNARE. With 28% amino acid identity to Bet1p, GS15 (Golgi SNARE of 15kDa) is widely expressed in rat tissues. It is an integral membrane protein of the Golgi apparatus and functions as a SNARE. Thus, GS15 is thought to be a novel SNARE that participates in ER-Golgi protein transport through an undefined process.
Immunofluorescence, Western Blotting
Specifications
| GS15 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| Golgi SNARE of 15 kDa |
| 19 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Frog, Human, Mouse, Rat | |
| Rat GS15 aa. 1-85 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.