Learn More
G protein-coupled receptor kinase 4, Mouse, Clone: 6D5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GRK4.
Brand: Abnova H00002868-M02.100ug
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating its deactivation. This gene has been linked to both genetic and acquired hypertension. [provided by RefSeq
Sequence: LPPVSQCSELRHSIEKDYSSLCDKQPIGRRLFRQFCDTKPTLKRHIEFLDAVAEYEVADDEDRSDCGLSILDRFFNDKLAAPLPEIPPDVVTECRLGLKEENPSKKAFEESpecifications
| G protein-coupled receptor kinase 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_182982 | |
| GRK4 | |
| GRK4 (NP_892027, 36 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6D5 | |
| Mouse monoclonal antibody raised against a partial recombinant GRK4. | |
| GRK4 | |
| GPRK2L/GPRK4/GRK4a/IT11 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2868 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.