missing translation for 'onlineSavingsMsg'
Learn More

GRB2 Mouse, Unlabeled, Clone: 81, BD

Mouse Monoclonal Antibody

£284.36 - £510.00

Specifications

Antigen GRB2
Clone 81
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15835269
View Documents
BD Biosciences
610111
50 μg
£510.00
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15845269
View Documents
BD Biosciences
610112
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Growth-factor Receptor-Bound Protein 2 (GRB2) was isolated by using the EGF receptor C-terminus as a probe in the screening of λgt11 expression libraries. This 24kDa GRB2 protein is ubiquitously expressed and consists of one SH2 domain flanked by two SH3 domains. Its ability to bind proteins through these domains has prompted much investigation of its role as an adaptor protein. Sos, a Ras GDP/GTP exchange protein, is constitutively bound to the SH3 domain of GRB2. Following growth factor stimulation, receptor tyrosine kinases autophosphorylate, creating a binding site for the GRB2 SH2 domain. Alternatively, GRB2 interacts with the receptor-bound active Shc protein.Through these interactions, the GRB2/Sos complex is translocated to the membrane where Sos activates membrane-bound Ras to initiate the Ras signaling pathway. Other proteins that similarly interact with GRB2 include Cbl, PTP1D, and Dynamin. However,the mechanisms through which these associations impact cellular responses remain to be discovered.

Host Species: Mouse
Clone: 81
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat GRB2 aa. 1-217
Formula Weight [Chemical]: 24kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

GRB2
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
81
Western Blot
Unconjugated
RUO
Canine, Chicken, Frog, Human, Mouse, Rat
Rat GRB2 aa. 1-217
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.