missing translation for 'onlineSavingsMsg'
Learn More

GRASP65 Polyclonal Antibody, Invitrogen™

GRASP65 Antibody, PA1-30604, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot.

Brand:  Invitrogen PA130604

Product Code. 11521443

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

GRASP65 Antibody, PA1-30604, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot.

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Isotype: IgG
Immunogen: Rabbits were immunized with recombinant (6His-tagged) mouse GRASP65 protein.
Formulation: 200μL unpurified antibody in whole serum with 50% glycerol and 0.01% thimerosal
Synonyms: Golgi reassembly-stacking protein of 65 kDa, Golgi phosphoprotein 5
Uniprot Accession: Q91X51
Entrez GeneID: 56082, 74498
Gene Symbol: GORASP1
PA1-30604 detects GRASP65 from rat, mouse samples. PA1-30604 has been successfully used in immunofluorescence, immunoprecipitation, Western blot applications. The PA1-30604 immunogen is: Rabbits were immunized with recombinant (6His-tagged) mouse GRASP65 protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRASP65
Polyclonal
Q91X51
GORASP1
Rabbits were immunized with recombinant (6His-tagged) mouse GRASP65 protein.
RUO
56082, 74498
Store at -20°C, Avoid Freeze/Thaw Cycles
IgG
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Golgi reassembly-stacking protein of 65 kDa, Golgi phosphoprotein 5
Rabbit
200 μL
Primary
Mouse, Rat
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.