Learn More
GRB2-related adaptor protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GRAP2 protein.
Brand: Abnova H00009402-D01P.100ug
Description
This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist. [provided by RefSeq
Sequence: MEAVAKFDFTASGEDELSFHTGDVLKILSNQEEWFKAELGSQEGYVPKNFIDIQFPKWFHEGLSRHQAENLLMGKEVGFFIIRASQSSPGDFSISVRHEDDVQHFKVMRDNKGNYFLWTEKFPSLNKLVDYYRTNSISRQKQIFLRDRTREDQGHRGNSLDRRSQGGPHLSGAVGEEIRPSMNRKLSDHPPTLPLQQHQHQPQPPQYAPAPQQLQQPPQQRYLQHHHFHQERRGGSLDINDGHCGTGLGSEMNAALMHRRHTDPVQLQAAGRVRWARALYDFEALEDDELGFHSGEVVEVLDSSNPSWWTGRLHNKLGLFPANYVAPMTRSpecifications
| GRB2-related adaptor protein 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human GRAP2 protein. | |
| GRAP2 | |
| GADS/GRAP-2/GRB2L/GRBLG/GRID/GRPL/GrbX/Grf40/Mona/P38 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004810 | |
| GRAP2 | |
| GRAP2 (NP_004801.1, 1 a.a. ∼ 330 a.a) full-length human protein. | |
| 100 μg | |
| Membrane Receptors | |
| 9402 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.