Learn More
Granzyme A Set Mouse anti-Human, FITC, Clone: CB9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558905
Description
The primary mechanism by which cytotoxic T cells eliminate virally infected cells is by granule exocytosis. The release of cytotoxic granule contents by cytotoxic T lymphocytes (CTL) triggers apoptotic target cell death. CTL granules contain a pore-forming protein, perforin, and a group of serine proteases called granzymes. In the classic model, perforins create holes in the target cell membrane, allowing entrance of the granzymes. Granzyme A and B are the predominant granzymes activated after CTL activation, but each act via an independent apoptotic pathway; granzyme B is activated immediately, while granzyme A acts hours later. Granzyme B does not induce cleavage of caspase-3, lamin B, rho-GTPase or PARP, but does cleave DNA-PKcs and nuclear mitotic apparatus protein (NuMA). The physiological substrates for granzyme A in the apoptotic pathway have not been identified. Studies involving mice which are deficient in both granzyme A and B suggest a model whereby the granzyme B pathway may have evolved as the major apoptotic pathway with the granzyme A pathway acting as a backup. However, further research is needed to delineate the components of the distinct pathways.
Clone CB9 recognizes human granzyme A. Purified human granzyme A was used as immunogen. Clone MOPC-21 is a mouse IgG1 isotype (negative) control. The MOPC-21 antibody has unknown specificity. CB9 and MOPC-21 FITC conjugates are optimized in F/P ratios.
The antibodies are routinely tested in parallel by flow cytometry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Granzyme A Set | |
| Monoclonal | |
| FITC | |
| Affinity Purified | |
| RUO | |
| FITC Human Granzyme A (100 tests), FITC Mouse IgG1, κ Isotype Control (100 tests) | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| CB9 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.