missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR91 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533787
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset, Dog, Bovine, Panda, Horse (88%).
This polyclonal antibody was raised in Rabbit against GPR91. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR91. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Specifications
| GPR91 | |
| Polyclonal | |
| Unconjugated | |
| SUCNR1 | |
| G protein-coupled receptor 91; GPR91; G-protein coupled receptor 91; G-protein-coupled receptor 91; p2u2; P2Y purinoceptor 1; P2Y purinoceptor 1-like; Succinate receptor; Succinate receptor 1; SUCNR1; SUCR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56670 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BXA5 | |
| SUCNR1 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GPR91. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction