Learn More
anti-GPR91, Polyclonal, Abcam™
Brand: Abcam AB75105-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human. Predicted to work with Mouse, Rat
Immunogen: Synthetic peptide (Human)(Peptide available as ab106670.)
Applications:
ELISA, Western Blotting
Specifications
| GPR91 | |
| 1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Signal Transduction, Tags And Cell Markers | |
| Human, Mouse, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide (Human)(Peptide available as ab106670.) | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.