Learn More
G protein-coupled receptor 77, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GPR77 protein.
Brand: Abnova H00027202-B01P.50ug
Description
The anaphylatoxins C3a (see MIM 120700), C4a (see MIM 120810), and C5a are cationic fragments generated during the complement cascade that participate in host defense. In the case of inappropriate complement activation, anaphylatoxins may be involved in autoimmunity and sepsis. C5L2 is coexpressed with the C5a receptor, C5AR (C5R1; MIM 113995), on polymorphonuclear neutrophils and may modulate C5AR activity (Gerard et al., 2005 [PubMed 16204243]).[supplied by OMIM
Sequence: MGNDSVSYEYGDYSDLSDRPVDCLDGACLAIDPLRVAPLPLYAAIFLVGVPGNAMVAWVAGKVARRRVGATWLLHLAVADLLCCLSLPILAVPIARGGHWPYGAVGCRALPSIILLTMYASVLLLAALSADLCFLALGPAWWSTVQRACGVQVACGAAWTLALLLTVPSAIYRRLHQEHFPARLQCVVDYGGSSSTENAVTAIRFLFGFLGPLVAVASCHSALLCWAARRCRPLGTAIVVGFFVCWAPYHLLGLVLTVAAPNSALLARALRAEPLIVGLALAHSCLNPMLFLYFGRAQLRRSLPAACHWALRESQGQDESVDSKKSTSHDLVSEMEVSpecifications
| G protein-coupled receptor 77 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human GPR77 protein. | |
| GPR77 | |
| C5L2/GPF77 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_018485.1 | |
| GPR77 | |
| GPR77 (NP_060955.1, 1 a.a. ∼ 337 a.a) full-length human protein. | |
| 50 μg | |
| Membrane Receptors | |
| 27202 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.