Learn More
anti-GPI, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00002821-AP22.1Set
Product Code. 16182104
  • £729.00 / 1 set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-GPI (100μg) , Detection antibody: mouse purified polyclonal anti-GPI (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
GPI
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GPI.
AMF/GNPI/NLK/PGI/PHI/SA-36
Mouse, Rabbit
RUO
2821
Human
Liquid
ELISA
Unconjugated
GPI
GPI
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-GPI (100 μg) 2. Detection antibody: mouse purified polyclonal anti-GPI (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.