missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-GPCR GPR119, Polyclonal, Abcam™
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse. Predicted to work with Rat
Immunogen: Synthetic peptide derived from C terminal domain of mouse GPCR GPR119 protein
Applications:
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | GPCR GPR119 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from C terminal domain of mouse GPCR GPR119 protein |
| Quantity | 100 μL |
| Research Discipline | Signal Transduction |
| Primary or Secondary | Primary |
| Target Species | Mouse, Rat |
| Form | Antisera |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?