Learn More
GP340 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522090
Description
PA5-22090 targets gp340 in WB applications and shows reactivity with Human samples. The PA5-22090 immunogen is a recombit fragment corresponding to a region within amino acids 2035 and 2313 of gp340 (Uniprot IDNo.Q9UGM3). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| gp340 | |
| Polyclonal | |
| Unconjugated | |
| Q9UGM3 | |
| DMBT1 | |
| Recombit fragment corresponding to a region within amino acids 2035 and 2313 of gp340 (Uniprot IDNo.Q9UGM3) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| DMBT1 | |
| RP11-481L19.1, GP340, muclin, SAG, deleted in maligt brain tumors 1 protein, glycoprotein 340, gp-340, hensin, salivary agglutinin, surfactant pulmonary-associated D-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.