Learn More
Golgin-84 Mouse, Unlabeled, Clone: 26, BD
Mouse Monoclonal Antibody
£428.00
Specifications
| Antigen | Golgin-84 |
|---|---|
| Clone | 26 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845909
|
BD Biosciences
611383 |
150 μg | |||||||
Description
The Golgi apparatus is a complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. Golgin-84 is an integral membrane protein associated with the Golgi. Sequence analysis of the C-terminal region of Golgin-84 demonstrates a 14 residue region extending into the lumen of the Golgi, a membrane insertion sequence, and a Golgi retention signal. The large N-terminal cytoplasmic region contains a coiled-coil domain that is required for dimerization and two leucine zipper domains. Golgin-84 is ubiquitously expressed, but is abundant in testis. Golgin-84 has sequence homology to coiled-coil containing myosin family members and to several other Golgi proteins, including trans-Golgi p230 and gigantin. Trans-Golgi p230 has been implicated in the biogenesis of specific Golgi vesicles, while gigantin is implicated in the tethering of non-clathrin coated vesicles and in the anchoring of adjacent Golgi cisternae. Thus, Golgin-84 may have similar roles in vesicle biogenesis, docking, and fusion at the Golgi interface.
Immunofluorescence, Western Blotting
Specifications
| Golgin-84 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 26 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human | |
| Human Golgin-84 aa. 510-713 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.