Learn More
golgi autoantigen, golgin subfamily a, 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GOLGA5 protein.
Brand: Abnova H00009950-D01P.100ug
Description
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins, whose members localize to the Golgi. This protein is a coiled-coil membrane protein that has been postulated to play a role in vesicle tethering and docking. Translocations involving this gene and the ret proto-oncogene have been found in tumor tissues; the chimeric sequences have been designated RET-II and PTC5. [provided by RefSeq
Sequence: MSWFVDLAGKAEDLLNRVDQGAATALSRKDNASNIYSKNTDYTELHQQNTDLIYQTGPKSTYISSAADNIRNQKATILAGTANVKVGSRTPVEASHPVENASVPRPSSHFVRRKKSEPDDELLFDFLNSSQKEPTGRVEIRKEKGKTPVFQSSQTSSVSSVNPSVTTIKTIEENSFGSQTHEAASNSDSSHEGQEESSKENVSSNAACPDHTPTPNDDGKSHELSNLRLENQLLRNEVQSLNQEMASLLQRSKETQEELNKARARVEKWNADHSKSDRMTRGLRAQVDDLTEAVAAKDSQLAVLKVRLQEADQLLSTRTEALEALQSEKSRIMQDQSEGNSLQNQALQTFQERLHEADATLKREQESYKQMQSEFAARLNKVEMERQNLAEAITLAERKYSDEKKRVDELQQQVKLYKLNLESSKQELIDYKQKATRILQSKEKLINSLKEGSGFEGLDSSTASSMELEELRHEKEMQREEIQKLMGQIHQLRSELQDMEAQQVNEAESAREQLQDLHDQIAGQKASKQELETELERLKQEFHYIEEDLYRTKNTLQSRIKDRDEEIQKLRNQLTNKTLSNSSQSELENRLHQLTETLIQKQTMLESLSTEKNSLVFQLERLEQQMNSASGSSSNGSSINMSGIDNGEGTRLRNVPVLFNDTETNLAGMYGKVRKAASSIDQFSIRLGIFLRRYPIARVFVIIYMALLHLWVMIVLLTYTPEMHHDQPYGKSpecifications
| golgi autoantigen, golgin subfamily a, 5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human GOLGA5 protein. | |
| GOLGA5 | |
| GOLIM5/RFG5/ret-II | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9950 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005113.2 | |
| GOLGA5 | |
| GOLGA5 (NP_005104.2, 1 a.a. ∼ 731 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.