Learn More
GOK/Stim1 Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610954
Description
The human chromosomal region 11p15 has undergone intense analysis because of its association with various malignancies. In particular, the band 11p15.5 contains genes associated with Wilms tumor, Beckwith-Weidemann syndrome, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. One such gene, GOK (Stim 1), was identified near the 5′ end of the ribonucleotide reductase subunit 1 gene. Examination of the GOK primary amino acid sequence indicates that it is a typical transmembrane protein with an extracellular N-terminal domain and a cytosolic C-terminal domain. The protein is highly hydrophobic with only a short region of hydrophobicity that likely represents the transmembrane region. The C-terminal portion of GOK shares some small regions of homology with myosin (20% identity). This region of GOK consists of α-helices and is thought to adopt a coiled-coil conformation. Although GOK expression has no effect on the growth of certain breast cancer cell lines, it induces death in rhabdomyosarcoma cells. Thus, it is thought to be a recessive tumor suppressor in muscle cells, possibly by functioning as a receptor connected to an apoptotic signaling pathway.
Immunofluorescence, Western Blotting
Specifications
| GOK/Stim1 | |
| Monoclonal | |
| 250μg/mL | |
| Stim1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 44 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human GOK aa. 25-139 | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.